4-Chloro-2-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]-5-piperazino-3(2H)-pyridazinone ≥95%

Supplier: Matrix Scientific

043854-500MG 043854-1G 043854-5G
101862-916EA 747.69 USD
101862-916 101862-918 101862-920
4-Chloro-2-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]-5-piperazino-3(2H)-pyridazinone ≥95%
4-Chloro-2-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]-5-piperazino-3(2H)-pyridazinone

Hazard: Irritant

Formula: C₁₄H₁₂Cl₂F₃N₅O
Melting Pt: 94…97 °C
MDL Number: MFCD00793846
CAS Number: 338399-95-4

Order Now

Specification Test Results

Melting point 94-97°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR