4-Chloro-2-[3-chloro-5-(trifluoromethyl)-2-pyridinyl]-5-[(2-hydroxyethyl)(methyl)amino]-3(2H)-pyridazinone ≥95%
Supplier: Matrix Scientific
043853-500MG
043853-1G
043853-5G
101862-910EA
747.69
USD
101862-910
101862-912
101862-914
4-Chloro-2-[3-chloro-5-(trifluoromethyl)-2-pyridinyl]-5-[(2-hydroxyethyl)(methyl)amino]-3(2H)-pyridazinone ≥95%
4-Chloro-2-[3-chloro-5-(trifluoromethyl)-2-pyridinyl]-5-[(2-hydroxyethyl)(methyl)amino]-3(2H)-pyridazinone
Hazard: Irritant
Formula:
C₁₃H₁₁Cl₂F₃N₄O₂ MW: 383.16 g/mol Melting Pt: 149…151 °C |
MDL Number:
MFCD00793844 CAS Number: 338399-91-0 |
Specification Test Results
Melting point | 149-151°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...