1-Allyl-2,4-dioxo-1,2,3,4-tetrahydro-5-pyrimidinecarbonitrile ≥95%

Supplier: Matrix Scientific

043851-500MG 043851-1G 043851-5G
101862-898EA 747.69 USD
101862-898 101862-900 101862-902
1-Allyl-2,4-dioxo-1,2,3,4-tetrahydro-5-pyrimidinecarbonitrile ≥95%
1-Allyl-2,4-dioxo-1,2,3,4-tetrahydro-5-pyrimidinecarbonitrile

Hazard: Irritant

Formula: C₈H₇N₃O₂
Melting Pt: 156…158 °C
MDL Number: MFCD01315928
CAS Number: 25855-27-0

Order Now

Specification Test Results

Melting point 156-158°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR