2-[(4-Methylphenyl)sulfanyl]nicotinaldehyde ≥95%
Supplier: Matrix Scientific
043850-500MG
043850-1G
043850-5G
101862-892EA
375.38
USD
101862-892
101862-894
101862-896
2-[(4-Methylphenyl)sulfanyl]nicotinaldehyde ≥95%
2-[(4-Methylphenyl)sulfanyl]nicotinaldehyde
Hazard: Irritant
Formula:
C₁₃H₁₁NOS Melting Pt: 82…84 °C |
MDL Number:
MFCD00794313 CAS Number: 338982-20-0 |
Specification Test Results
Melting point | 82-84°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...