(2-Chloro-3-pyridinyl)methanol ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Chloro-3-pyridinemethanol, (2-chloropyridin-3-yl)methanol, 2-Chloro-3-pyridinylmethanol

043848-1G 043848-5G
101862-888EA 15.38 USD
101862-888 101862-890
(2-Chloro-3-pyridinyl)methanol ≥95%
(2-Chloro-3-pyridinyl)methanol

Hazard: Irritant

Formula: C₆H₆ClNO
MW: 143.57 g/mol
Melting Pt: 62…64 °C
MDL Number: MFCD01935961
CAS Number: 42330-59-6

Order Now

Specification Test Results

Melting point 62-64°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR