3-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]sulfanyl}-5-methyl-4-isothiazolecarbonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 3-{[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]thio}-4-cyano-5-methylisothiazole

043847-500MG 043847-1G 043847-5G
101862-882EA 556.92 USD
101862-882 101862-884 101862-886
3-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]sulfanyl}-5-methyl-4-isothiazolecarbonitrile ≥95%
3-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]sulfanyl}-5-methyl-4-isothiazolecarbonitrile

Hazard: Irritant

Formula: C₁₁H₅ClF₃N₃S₂
MW: 335.76 g/mol
Melting Pt: 122…125 °C
MDL Number: MFCD00244313
CAS Number: 338775-63-6

Order Now

Specification Test Results

Melting point 122-125°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR