3-(2-Hydroxyphenoxy)-2-butanone ≥95%

Supplier: Matrix Scientific

043846-500MG 043846-1G 043846-5G
101862-876EA 181.54 USD
101862-876 101862-878 101862-880
3-(2-Hydroxyphenoxy)-2-butanone ≥95%
3-(2-Hydroxyphenoxy)-2-butanone

Hazard: Irritant

Formula: C₁₀H₁₂O₃
MW: 180.21 g/mol
Melting Pt: 70…72 °C
MDL Number: MFCD00664263
CAS Number: 91061-42-6

Order Now

Specification Test Results

Melting point 70-72°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR