5-Chloro-4-(chloromethyl)-1-methyl-3-phenyl-1H-pyrazole ≥95%

Supplier: Matrix Scientific

043844-500MG 043844-1G 043844-5G
101862-864EA 556.92 USD
101862-864 101862-866 101862-868
5-Chloro-4-(chloromethyl)-1-methyl-3-phenyl-1H-pyrazole ≥95%
5-Chloro-4-(chloromethyl)-1-methyl-3-phenyl-1H-pyrazole

Hazard: Irritant

Formula: C₁₁H₁₀Cl₂N₂
Melting Pt: 70…74 °C
MDL Number: MFCD00244901
CAS Number: 321538-19-6

Order Now

Specification Test Results

Melting point 70-74°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR