(5-Chloro-1-methyl-3-phenyl-1H-pyrazol-4-yl)methanol ≥95%

Supplier: Matrix Scientific

043843-500MG 043843-1G 043843-5G
101862-858EA 455.38 USD
101862-858 101862-860 101862-862
(5-Chloro-1-methyl-3-phenyl-1H-pyrazol-4-yl)methanol ≥95%
(5-Chloro-1-methyl-3-phenyl-1H-pyrazol-4-yl)methanol

Hazard: Irritant

Formula: C₁₁H₁₁ClN₂O
Melting Pt: 130…133 °C
MDL Number: MFCD00244900
CAS Number: 321538-17-4

Order Now

Specification Test Results

Melting point 130-133°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR