6-Methyl-2-oxo-4-(trifluoromethyl)-1,2-dihydro-3-pyridinecarbonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 6-Methyl-2-oxo-4-(trifluoromethyl)-1,2-dihydropyridine-3-carbonitrile

043841-500MG 043841-1G 043841-5G
101862-846EA 181.54 USD
101862-846 101862-848 101862-850
6-Methyl-2-oxo-4-(trifluoromethyl)-1,2-dihydro-3-pyridinecarbonitrile ≥95%
6-Methyl-2-oxo-4-(trifluoromethyl)-1,2-dihydro-3-pyridinecarbonitrile

Hazard: Irritant

Formula: C₈H₅F₃N₂O
MW: 202.14 g/mol
Melting Pt: 232…234 °C
MDL Number: MFCD01314908
CAS Number: 654-49-9

Order Now

Specification Test Results

Melting point 232-234°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR