8-Chloro-6-(trifluoromethyl)[1,2,4]triazolo[4,3-a]pyridin-3-amine ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Amino-8-chloro-6-(trifluoromethyl)[1,2,4]triazolo[4,3-a]pyridine

043840-500MG 043840-1G
101862-842EA 921.54 USD
101862-842 101862-844
8-Chloro-6-(trifluoromethyl)[1,2,4]triazolo[4,3-a]pyridin-3-amine ≥95%
8-Chloro-6-(trifluoromethyl)[1,2,4]triazolo[4,3-a]pyridin-3-amine

Hazard: Irritant

Formula: C₇H₄ClF₃N₄
Melting Pt: 273…275 °C
MDL Number: MFCD00974657
CAS Number: 338773-34-5

Order Now

Specification Test Results

Melting point 273-275°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR