4-Methoxy-2-oxo-1,2-dihydropyridine-3-carbonitrile ≥95%
Supplier: Matrix Scientific
Synonyms:
N-Demethylricinine
043839-1G
043839-5G
043839-25G
101862-838EA
100
USD
101862-838
101862-840
102551-328
4-Methoxy-2-oxo-1,2-dihydropyridine-3-carbonitrile ≥95%
4-Methoxy-2-oxo-1,2-dihydropyridine-3-carbonitrile
Hazard: Irritant
Formula:
C₇H₆N₂O₂ MW: 150.13 g/mol Melting Pt: 293 °C |
MDL Number:
MFCD00975442 CAS Number: 21642-98-8 |
Specification Test Results
Melting point | 293°C |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...