2-Amino-4-methoxynicotinonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Amino-4-methoxy-3-pyridinecarbonitrile

043838-500MG 043838-1G 043838-5G
101862-832EA 109.23 USD
101862-832 101862-834 101862-836
2-Amino-4-methoxynicotinonitrile ≥95%
2-Amino-4-methoxynicotinonitrile

Hazard: Irritant

Formula: C₇H₇N₃O
MW: 149.1500 g/mol
Melting Pt: 211°C
MDL Number: MFCD00975441
CAS Number: 98651-70-8

Order Now

Specification Test Results

Melting point 211°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR