4-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]oxy}-2-buten-1-ol ≥95%

Supplier: Matrix Scientific

043836-500MG 043836-1G 043836-5G
101862-820EA 375.38 USD
101862-820 101862-822 101862-824
4-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]oxy}-2-buten-1-ol ≥95%
4-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]oxy}-2-buten-1-ol

Hazard: Irritant

Formula: C₁₀H₉ClF₃NO₂
MW: 267.64 g/mol
Melting Pt: 50…52 °C
MDL Number: MFCD04038247
CAS Number: 338772-77-3

Order Now

Specification Test Results

Melting point 50-52°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR