3-Methyl-3,4-dihydro-2(1H)-quinoxalinone ≥95%

Supplier: Matrix Scientific

043835-500MG 043835-1G 043835-5G
101862-814EA 78.46 USD
101862-814 101862-816 101862-818
3-Methyl-3,4-dihydro-2(1H)-quinoxalinone ≥95%
3-Methyl-3,4-dihydro-2(1H)-quinoxalinone

Hazard: Irritant

Formula: C₉H₁₀N₂O
MW: 162.19 g/mol
Melting Pt: 138…141 °C
MDL Number: MFCD00231317
CAS Number: 34070-68-3

Order Now

Specification Test Results

Melting point 138-141°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR