Ethyl 2-cyano-3-(4-methoxyphenyl)acrylate ≥95%

Supplier: Matrix Scientific

043834-500MG 043834-1G 043834-5G
101862-808EA 181.54 USD
101862-808 101862-810 101862-812
Ethyl 2-cyano-3-(4-methoxyphenyl)acrylate ≥95%
Ethyl 2-cyano-3-(4-methoxyphenyl)acrylate

Hazard: Irritant

Formula: C₁₃H₁₃NO₃
Melting Pt: 85…87 °C
MDL Number: MFCD00026849
CAS Number: 2017-87-0

Order Now

Specification Test Results

Melting point 85-87°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR