1-(3-Chloro-5-(trifluoromethyl)-2-pyridinyl)-1H-indole-3-carbaldehyde ≥95%

Supplier: Matrix Scientific

043831-500MG 043831-1G 043831-5G
101862-790EA 747.69 USD
101862-790 101862-792 101862-794
1-(3-Chloro-5-(trifluoromethyl)-2-pyridinyl)-1H-indole-3-carbaldehyde ≥95%
1-(3-Chloro-5-(trifluoromethyl)-2-pyridinyl)-1H-indole-3-carbaldehyde

Hazard: Irritant

Formula: C₁₅H₈ClF₃N₂O
Melting Pt: 132…134 °C
MDL Number: MFCD00664496
CAS Number: 338770-32-4

Order Now

Specification Test Results

Melting point 132-134°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR