1-(4-Chlorobenzyl)-2,4-dioxo-1,2,3,4-tetrahydro-5-pyrimidinecarbonitrile ≥95%

Supplier: Matrix Scientific

043828-500MG 043828-1G 043828-5G
101862-772EA 747.69 USD
101862-772 101862-774 101862-776
1-(4-Chlorobenzyl)-2,4-dioxo-1,2,3,4-tetrahydro-5-pyrimidinecarbonitrile ≥95%
1-(4-Chlorobenzyl)-2,4-dioxo-1,2,3,4-tetrahydro-5-pyrimidinecarbonitrile

Hazard: Irritant

Formula: C₁₂H₈ClN₃O₂
Melting Pt: 210…212 °C
MDL Number: MFCD00172960
CAS Number: 338399-17-0

Order Now

Specification Test Results

Melting point 210-212°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR