4-({[3-(Trifluoromethyl)phenyl]sulfanyl}methyl)aniline ≥95%

Supplier: Matrix Scientific

Synonyms: 3-(4-Aminobenzylthio)benzotrifluoride

043824-500MG 043824-1G 043824-5G
101862-748EA 921.54 USD
101862-748 101862-750 101862-752
4-({[3-(Trifluoromethyl)phenyl]sulfanyl}methyl)aniline ≥95%
4-({[3-(Trifluoromethyl)phenyl]sulfanyl}methyl)aniline

Hazard: Irritant

Formula: C₁₄H₁₂F₃NS
MW: 283.32 g/mol
Melting Pt: 68…71 °C
MDL Number: MFCD00173208
CAS Number: 338398-58-6

Order Now

Specification Test Results

Melting point 68-71°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR