2,2,2-Trichloro-1-{4-[2-(4-chlorophenyl)acetyl]-1H-pyrrol-2-yl}-1-ethanone ≥95%

Supplier: Matrix Scientific

043820-500MG 043820-1G 043820-5G
101862-724EA 556.92 USD
101862-724 101862-726 101862-728
2,2,2-Trichloro-1-{4-[2-(4-chlorophenyl)acetyl]-1H-pyrrol-2-yl}-1-ethanone ≥95%
2,2,2-Trichloro-1-{4-[2-(4-chlorophenyl)acetyl]-1H-pyrrol-2-yl}-1-ethanone

Hazard: Irritant

Formula: C₁₄H₉Cl₄NO₂
Melting Pt: 147…150 °C
MDL Number: MFCD00173091
CAS Number: 338397-93-6

Order Now

Specification Test Results

Melting point 147-150°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR