1-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-1H-pyrrole-2-sulfonoyl chloride ≥95%

Supplier: Matrix Scientific

043819-500MG 043819-1G 043819-5G
101862-718EA 426.15 USD
101862-718 101862-720 101862-722
1-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-1H-pyrrole-2-sulfonoyl chloride ≥95%
1-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-1H-pyrrole-2-sulfonoyl chloride

Hazard: Irritant

Formula: C₁₀H₅Cl₂F₃N₂O₂S
Melting Pt: 71…74 °C
MDL Number: MFCD00214861
CAS Number: 338397-79-8

Order Now

Specification Test Results

Melting point 71-74°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR