N-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-3-(hydrazinocarbonyl)-5-methyl-4-isoxazolecarboxamide ≥95%

Supplier: Matrix Scientific

043818-500MG 043818-1G
101862-714EA 1855.38 USD
101862-714 101862-716
N-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-3-(hydrazinocarbonyl)-5-methyl-4-isoxazolecarboxamide ≥95%
N-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-3-(hydrazinocarbonyl)-5-methyl-4-isoxazolecarboxamide

Hazard: Irritant

Formula: C₁₂H₉ClF₃N₅O₃
MW: 363.69 g/mol
Melting Pt: 146…148 °C
MDL Number: MFCD00664740
CAS Number: 338397-37-8

Order Now

Specification Test Results

Melting point 146-148°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR