Ethyl-2-(3,4-dimethoxyphenyl)-1,3-thiazole-4-carboxylate ≥95%
Supplier: Matrix Scientific
043815-500MG
043815-1G
043815-5G
101862-702EA
400
USD
101862-702
101862-704
101862-706
Ethyl-2-(3,4-dimethoxyphenyl)-1,3-thiazole-4-carboxylate ≥95%
Ethyl-2-(3,4-dimethoxyphenyl)-1,3-thiazole-4-carboxylate
Hazard: Irritant
Formula:
C₁₄H₁₅NO₄S Melting Pt: 96…99 °C |
MDL Number:
MFCD00665469 CAS Number: 338982-18-6 |
Specification Test Results
Melting point | 96-99°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...