2-(4-Methylphenyl)-4-thiazolecarboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 2-(p-Tolyl)thiazole-4-carboxylic acid

043813-500MG 043813-1G 043813-5G
101862-690EA 747.69 USD
101862-690 101862-692 101862-694
2-(4-Methylphenyl)-4-thiazolecarboxylic acid ≥95%
2-(4-Methylphenyl)-4-thiazolecarboxylic acid

Hazard: Irritant

Formula: C₁₁H₉NO₂S
MW: 219.26 g/mol
Melting Pt: 176-178°C
MDL Number: MFCD01936002
CAS Number: 17228-99-8

Order Now

Specification Test Results

Melting point 176-178°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR