Ethyl-2-(4-methylphenyl)-1,3-thiazole-4-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 2-(p-tolyl)thiazole-4-carboxylate

043812-500MG 043812-1G 043812-5G
101862-684EA 375.38 USD
101862-684 101862-686 101862-688
Ethyl-2-(4-methylphenyl)-1,3-thiazole-4-carboxylate ≥95%
Ethyl-2-(4-methylphenyl)-1,3-thiazole-4-carboxylate

Hazard: Irritant

Formula: C₁₃H₁₃NO₂S
MW: 247.32 g/mol
Melting Pt: 45…46 °C
MDL Number: MFCD01935979
CAS Number: 132089-32-8

Order Now

Specification Test Results

Melting point 45-46°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR