N-(4-Chloro-6-methoxy-2-pyrimidinyl)-N,N-dimethylamine ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Chloro-6-methoxy-N,N-dimethylpyrimidin-2-amine

043811-500MG 043811-1G 043811-5G
101862-678EA 747.69 USD
101862-678 101862-680 101862-682
N-(4-Chloro-6-methoxy-2-pyrimidinyl)-N,N-dimethylamine ≥95%
N-(4-Chloro-6-methoxy-2-pyrimidinyl)-N,N-dimethylamine

Hazard: Irritant

Formula: C₇H₁₀ClN₃O
MW: 187.63 g/mol
Melting Pt: 139…142 °C
MDL Number: MFCD01935946
CAS Number: 13012-26-5

Order Now

Specification Test Results

Melting point 139-142°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR