4-Methyl-2-(4-methylphenyl)-1,3-thiazole-5-carboxylic acid ≥95%
Supplier: Matrix Scientific
043809-500MG
043809-1G
043809-5G
101862-666EA
747.69
USD
101862-666
101862-668
101862-670
4-Methyl-2-(4-methylphenyl)-1,3-thiazole-5-carboxylic acid ≥95%
4-Methyl-2-(4-methylphenyl)-1,3-thiazole-5-carboxylic acid
Hazard: Irritant
Formula:
C₁₂H₁₁NO₂S Melting Pt: 262…263.5 °C |
MDL Number:
MFCD01935999 CAS Number: 54001-13-7 |
Specification Test Results
Melting point | 262-263.5°C |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...