4,6-Dichloro-N,N-dimethyl-2-pyrimidinamine ≥95%

Supplier: Matrix Scientific

043808-500MG 043808-1G 043808-5G
101862-660EA 375.38 USD
101862-660 101862-662 101862-664
4,6-Dichloro-N,N-dimethyl-2-pyrimidinamine ≥95%
4,6-Dichloro-N,N-dimethyl-2-pyrimidinamine

Hazard: Irritant

Formula: C₆H₇Cl₂N₃
MW: 192.05 g/mol
Melting Pt: 56…57.5 °C
MDL Number: MFCD01935962
CAS Number: 5734-68-9

Order Now

Specification Test Results

Melting point 56-57.5°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR