4-Chloro-3-nitrobenzenecarbaldehyde oxime ≥95%

Supplier: Matrix Scientific

043805-500MG 043805-1G 043805-5G
101862-648EA 278.46 USD
101862-648 101862-650 101862-652
4-Chloro-3-nitrobenzenecarbaldehyde oxime ≥95%
4-Chloro-3-nitrobenzenecarbaldehyde oxime

Hazard: Irritant

Formula: C₇H₅ClN₂O₃
Melting Pt: 147…150 °C
MDL Number: MFCD00160759
CAS Number: 66399-01-7

Order Now

Specification Test Results

Melting point 147-150°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR