3-(Dimethylamino)-2-[3-(trifluoromethyl)phenyl]acrylonitrile ≥95%

Supplier: Matrix Scientific

043802-500MG 043802-1G 043802-5G
101862-636EA 181.54 USD
101862-636 101862-638 101862-640
3-(Dimethylamino)-2-[3-(trifluoromethyl)phenyl]acrylonitrile ≥95%
3-(Dimethylamino)-2-[3-(trifluoromethyl)phenyl]acrylonitrile

Hazard: Irritant

Formula: C₁₂H₁₁F₃N₂
Melting Pt: 104…107 °C
MDL Number: MFCD00215017
CAS Number: 62738-99-2

Order Now

Specification Test Results

Melting point 104-107°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR