3-Imino-3-(4-phenylpiperazino)propanenitrile ≥95%

Supplier: Matrix Scientific

043800-500MG 043800-1G 043800-5G
101862-626EA 747.69 USD
101862-626 101862-628 101862-630
3-Imino-3-(4-phenylpiperazino)propanenitrile ≥95%
3-Imino-3-(4-phenylpiperazino)propanenitrile

Hazard: Irritant

Formula: C₁₃H₁₆N₄
Melting Pt: 182…184 °C
MDL Number: MFCD00664737
CAS Number: 338396-67-1

Order Now

Specification Test Results

Melting point 182-184°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR