5-(2-Chlorobenzylthio)-1-methyl-3-(trifluoromethyl)-1H-pyrazole-4-carboxaldehyde ≥95%

Supplier: Matrix Scientific

043797-500MG 043797-1G 043797-5G
101862-608EA 747.69 USD
101862-608 101862-610 101862-612
5-(2-Chlorobenzylthio)-1-methyl-3-(trifluoromethyl)-1H-pyrazole-4-carboxaldehyde ≥95%
5-(2-Chlorobenzylthio)-1-methyl-3-(trifluoromethyl)-1H-pyrazole-4-carboxaldehyde

Hazard: Irritant

Formula: C₁₃H₁₀ClF₃N₂OS
Melting Pt: 86…88 °C
MDL Number: MFCD00140828
CAS Number: 321533-90-8

Order Now

Specification Test Results

Melting point 86-88°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR