1-Methyl-5-(phenylsulfanyl)-3-(trifluoromethyl)-1H-pyrazole-4-carbaldehyde ≥95%

Supplier: Matrix Scientific

043796-500MG 043796-1G 043796-5G
101862-602EA 556.92 USD
101862-602 101862-604 101862-606
1-Methyl-5-(phenylsulfanyl)-3-(trifluoromethyl)-1H-pyrazole-4-carbaldehyde ≥95%
1-Methyl-5-(phenylsulfanyl)-3-(trifluoromethyl)-1H-pyrazole-4-carbaldehyde

Hazard: Irritant

Formula: C₁₂H₉F₃N₂OS
MW: 286.28 g/mol
Melting Pt: 96 °C
MDL Number: MFCD00140823
CAS Number: 321533-77-1

Order Now

Specification Test Results

Melting point 96°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR