5-[(4-Chlorophenyl)sulfanyl]-1-methyl-3-(trifluoromethyl)-1H-pyrazole-4-carbaldehyde ≥95%

Supplier: Matrix Scientific

043795-500MG 043795-1G 043795-5G
101862-596EA 747.69 USD
101862-596 101862-598 101862-600
5-[(4-Chlorophenyl)sulfanyl]-1-methyl-3-(trifluoromethyl)-1H-pyrazole-4-carbaldehyde ≥95%
5-[(4-Chlorophenyl)sulfanyl]-1-methyl-3-(trifluoromethyl)-1H-pyrazole-4-carbaldehyde

Hazard: Irritant

Formula: C₁₂H₈ClF₃N₂OS
MW: 320.72 g/mol
Melting Pt: 93…95 °C
MDL Number: MFCD00140822
CAS Number: 321533-70-4

Order Now

Specification Test Results

Melting point 93-95°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR