5-Chloro-1-methyl-3-(trifluoromethyl)-1H-pyrazole-4-carboxylic acid ≥95%

Supplier: Matrix Scientific

043794-500MG 043794-1G 043794-5G
101862-590EA 747.69 USD
101862-590 101862-592 101862-594
5-Chloro-1-methyl-3-(trifluoromethyl)-1H-pyrazole-4-carboxylic acid ≥95%
5-Chloro-1-methyl-3-(trifluoromethyl)-1H-pyrazole-4-carboxylic acid

Hazard: Irritant

Formula: C₆H₄ClF₃N₂O₂
Melting Pt: 200…202 °C
MDL Number: MFCD00215443
CAS Number: 128455-63-0

Order Now

Specification Test Results

Melting point 200-202°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR