6-(1H-Imidazol-1-yl)nicotinic acid ≥95%

Supplier: Matrix Scientific

043793-500MG 043793-1G 043793-5G
101862-584EA 278.46 USD
101862-584 101862-586 101862-588
6-(1H-Imidazol-1-yl)nicotinic acid ≥95%
6-(1H-Imidazol-1-yl)nicotinic acid

Hazard: Irritant

Formula: C₉H₇N₃O₂
Melting Pt: >270 °C
MDL Number: MFCD00140821
CAS Number: 216955-75-8

Order Now

Specification Test Results

Melting point >270°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR