4-Fluorophenyloxamic acid ethyl ester ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 2-((4-fluorophenyl)amino)-2-oxoacetate, Ethyl 2-(4-fluoroanilino)-2-oxoacetate

043787-500MG 043787-1G 043787-5G
101862-560EA 181.54 USD
101862-560 101862-562 101862-564
4-Fluorophenyloxamic acid ethyl ester ≥95%
4-Fluorophenyloxamic acid ethyl ester

Hazard: Irritant

Formula: C₁₀H₁₀FNO₃
MW: 211.1897 g/mol
Melting Pt: 120-122°C
MDL Number: MFCD00119045
CAS Number: 69065-91-4

Order Now

Specification Test Results

Melting point 120-122°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR