Methyl-4-chloro-1-(4-methoxyphenyl)-6-oxo-1,6-dihydro-3-pyridazinecarboxylate ≥95%

Supplier: Matrix Scientific

043786-500MG 043786-1G 043786-5G
101862-554EA 747.69 USD
101862-554 101862-556 101862-558
Methyl-4-chloro-1-(4-methoxyphenyl)-6-oxo-1,6-dihydro-3-pyridazinecarboxylate ≥95%
Methyl-4-chloro-1-(4-methoxyphenyl)-6-oxo-1,6-dihydro-3-pyridazinecarboxylate

Hazard: Irritant

Formula: C₁₃H₁₁ClN₂O₄
Melting Pt: 140…142 °C
MDL Number: MFCD00141156
CAS Number: 338395-95-2

Order Now

Specification Test Results

Melting point 140-142°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR