2-{[1-Methyl-5-(2,2,2-trichloroacetyl)-1H-pyrrol-3-yl]carbonyl}benzenecarboxylic acid ≥95%

Supplier: Matrix Scientific

043785-500MG 043785-1G 043785-5G
101862-548EA 556.92 USD
101862-548 101862-550 101862-552
2-{[1-Methyl-5-(2,2,2-trichloroacetyl)-1H-pyrrol-3-yl]carbonyl}benzenecarboxylic acid ≥95%
2-{[1-Methyl-5-(2,2,2-trichloroacetyl)-1H-pyrrol-3-yl]carbonyl}benzenecarboxylic acid

Hazard: Irritant

Formula: C₁₅H₁₀Cl₃NO₄
Melting Pt: 214…216 °C
MDL Number: MFCD00141858
CAS Number: 338395-82-7

Order Now

Specification Test Results

Melting point 214-216°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR