Methyl-4-chloro-6-oxo-1-[3-(trifluoromethyl)phenyl]-1,6-dihydro-3-pyridazinecarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 4-chloro-1,6-dihydro-6-oxo-1-[3-(trifluoromethyl)phenyl]pyridazine-3-carboxylate

043782-500MG 043782-1G
101862-532EA 747.69 USD
101862-532 101862-534
Methyl-4-chloro-6-oxo-1-[3-(trifluoromethyl)phenyl]-1,6-dihydro-3-pyridazinecarboxylate ≥95%
Methyl-4-chloro-6-oxo-1-[3-(trifluoromethyl)phenyl]-1,6-dihydro-3-pyridazinecarboxylate

Hazard: Irritant

Formula: C₁₃H₈ClF₃N₂O₃
MW: 332.67 g/mol
Melting Pt: 98…100 °C
MDL Number: MFCD00141148
CAS Number: 129109-18-8

Order Now

Specification Test Results

Melting point 98-100°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR