1-Methyl-1H-pyrrole-2-carbohydrazide ≥95%

Supplier: Matrix Scientific

043780-500MG 043780-1G 043780-5G
101862-520EA 375.38 USD
101862-520 101862-522 101862-524
1-Methyl-1H-pyrrole-2-carbohydrazide ≥95%
1-Methyl-1H-pyrrole-2-carbohydrazide

Hazard: Irritant

Formula: C₆H₉N₃O
Melting Pt: 126…128 °C
MDL Number: MFCD00141854
CAS Number: 113398-02-0

Order Now

Specification Test Results

Melting point 126-128°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR