Methyl-2-(benzoylamino)-3-(dimethylamino)acrylate ≥95%

Supplier: Matrix Scientific

043778-500MG 043778-1G 043778-5G
101862-508EA 181.54 USD
101862-508 101862-510 101862-512
Methyl-2-(benzoylamino)-3-(dimethylamino)acrylate ≥95%
Methyl-2-(benzoylamino)-3-(dimethylamino)acrylate

Hazard: Irritant

Formula: C₁₃H₁₆N₂O₃
Melting Pt: 170…172 °C
MDL Number: MFCD00202513
CAS Number: 56952-04-6

Order Now

Specification Test Results

Melting point 170-172°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR