2-[3-(Trifluoromethyl)phenoxy]ethanethioamide ≥95%

Supplier: Matrix Scientific

043776-500MG 043776-1G 043776-5G
101862-502EA 375.38 USD
101862-502 101862-504 101862-506
2-[3-(Trifluoromethyl)phenoxy]ethanethioamide ≥95%
2-[3-(Trifluoromethyl)phenoxy]ethanethioamide

Hazard: Irritant

Formula: C₉H₈F₃NOS
Melting Pt: 140…142 °C
MDL Number: MFCD00172129
CAS Number: 20293-32-7

Order Now

Specification Test Results

Melting point 140-142°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR