2,2,2-Trichloro-1-[4-(2-phenylacetyl)-1H-pyrrol-2-yl]-1-ethanone ≥95%

Supplier: Matrix Scientific

043773-500MG 043773-1G 043773-5G
101862-490EA 556.92 USD
101862-490 101862-492 101862-494
2,2,2-Trichloro-1-[4-(2-phenylacetyl)-1H-pyrrol-2-yl]-1-ethanone ≥95%
2,2,2-Trichloro-1-[4-(2-phenylacetyl)-1H-pyrrol-2-yl]-1-ethanone

Hazard: Irritant

Formula: C₁₄H₁₀Cl₃NO₂
Melting Pt: 169…171 °C
MDL Number: MFCD00141852
CAS Number: 338394-85-7

Order Now

Specification Test Results

Melting point 169-171°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR