2-Cyano-4'-methoxyacetanilide ≥95%
Supplier: Matrix Scientific
043770-500MG
043770-1G
043770-5G
101862-478EA
181.54
USD
101862-478
101862-480
101862-482
2-Cyano-4'-methoxyacetanilide ≥95%
2-Cyano-4'-methoxyacetanilide
Hazard: Irritant
Formula:
C₁₀H₁₀N₂O₂ Melting Pt: 133-135°C |
MDL Number:
MFCD00214717 CAS Number: 5382-38-7 |
Specification Test Results
Melting point | 133-135°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...