2-(2-Quinoxalinylsulfanyl)acetic acid ≥95%

Supplier: Matrix Scientific

043769-500MG 043769-1G 043769-5G
101862-472EA 110.77 USD
101862-472 101862-474 101862-476
2-(2-Quinoxalinylsulfanyl)acetic acid ≥95%
2-(2-Quinoxalinylsulfanyl)acetic acid

Hazard: Irritant

Formula: C₁₀H₈N₂O₂S
Melting Pt: 157…159 °C
MDL Number: MFCD00138691
CAS Number: 69001-90-7

Order Now

Specification Test Results

Melting point 157-159°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR