4-(3-Isopropenyl-2-oxo-2,3-dihydro-1H-1,3-benzimidazol-1-yl)butanoic acid ≥95%
Supplier: Matrix Scientific
043765-500MG
043765-1G
043765-5G
101862-462EA
375.38
USD
101862-462
101862-464
101862-466
4-(3-Isopropenyl-2-oxo-2,3-dihydro-1H-1,3-benzimidazol-1-yl)butanoic acid ≥95%
4-(3-Isopropenyl-2-oxo-2,3-dihydro-1H-1,3-benzimidazol-1-yl)butanoic acid
Hazard: Irritant
Formula:
C₁₄H₁₆N₂O₃ Melting Pt: 109…111 °C |
MDL Number:
MFCD08056613 CAS Number: 52099-78-2 |
Specification Test Results
Melting point | 109-111°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...