Ethyl 2-chloro-2-(2-(p-tolyl)hydrazono)acetate ≥95%

Supplier: Matrix Scientific

043762-500MG 043762-1G 043762-5G
101862-446EA 124.62 USD
101862-446 101862-448 101862-450
Ethyl 2-chloro-2-(2-(p-tolyl)hydrazono)acetate ≥95%
Ethyl 2-chloro-2-(2-(p-tolyl)hydrazono)acetate

Hazard: Irritant

Formula: C₁₁H₁₃ClN₂O₂
Melting Pt: 103…105 °C
MDL Number: MFCD02065467
CAS Number: 27171-88-6

Order Now

Specification Test Results

Melting point 103-105°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR