2-Amino-5-(3-fluorophenyl)-4-thiazolecarbohydrazide ≥95%

Supplier: Matrix Scientific

043759-500MG 043759-1G 043759-5G
101862-428EA 278.46 USD
101862-428 101862-430 101862-432
2-Amino-5-(3-fluorophenyl)-4-thiazolecarbohydrazide ≥95%
2-Amino-5-(3-fluorophenyl)-4-thiazolecarbohydrazide

Hazard: Irritant

Formula: C₁₀H₉FN₄OS
Melting Pt: 122°C
MDL Number: MFCD06200940
CAS Number: 886361-60-0

Order Now

Specification Test Results

Melting point 122°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR