2-Amino-5-(4-fluorophenyl)-4-thiazolecarbohydrazide ≥95%

Supplier: Matrix Scientific

043758-500MG 043758-1G 043758-5G
101862-422EA 278.46 USD
101862-422 101862-424 101862-426
2-Amino-5-(4-fluorophenyl)-4-thiazolecarbohydrazide ≥95%
2-Amino-5-(4-fluorophenyl)-4-thiazolecarbohydrazide

Hazard: Irritant

Formula: C₁₀H₉FN₄OS
Melting Pt: 174°C
MDL Number: MFCD06200939
CAS Number: 886361-58-6

Order Now

Specification Test Results

Melting point 174°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR